Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030384.1 | NP_001025555.1 | 8 - 9 | 1565 | 112 | |
XM_012961211.2 | XP_012816665.1 | 9 - 10 | 1950 | 112 | ||
XM_012961213.2 | XP_012816667.1 | 8 - 9 | 1572 | 112 | ||
GenBank mRNA | BC090595.1 | - | 8 - 9 | 1565 | 112 | |
CR760909.2 | - | 8 - 9 | 1561 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |