Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030419.1 | NP_001025590.1 | 2 - 3 | 961 | 81 | |
GenBank mRNA | BC091039.1 | - | 2 - 3 | 961 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |