Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032353.1 | NP_001027524.1 | 2 - 3 | 1158 | 110 | |
GenBank mRNA | BC099621.1 | - | 2 - 3 | 1158 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |