Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011087.2 | NP_001011087.2 | 2 - 3 | 385 | 70 | |
GenBank mRNA | CR761105.1 | - | 2 - 3 | 385 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |