Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016259.3 | NP_001016259.1 | 4 - 5 | 811 | 66 | |
XM_012968378.2 | XP_012823832.1 | 5 - 6 | 943 | 66 | ||
XM_012968380.2 | XP_012823834.1 | 4 - 5 | 718 | 66 | ||
XM_018095391.1 | XP_017950880.1 | 5 - 6 | 984 | 66 | ||
GenBank mRNA | CR761716.2 | - | 4 - 5 | 812 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |