Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016433.2 | NP_001016433.1 | 2 - 3 | 736 | 64 | |
XM_012968410.2 | XP_012823864.1 | 2 - 3 | 902 | 64 | ||
GenBank mRNA | BC170942.1 | - | 2 - 3 | 693 | 64 | |
BC170944.1 | - | 2 - 3 | 693 | 64 | ||
CR761045.2 | - | 2 - 3 | 736 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |