Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016617.2 | NP_001016617.1 | 3 - 4 | 237 | 70 | |
GenBank mRNA | BC118689.1 | - | 3 - 4 | 201 | 70 | |
CR760508.2 | - | 3 - 4 | 237 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |