Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsmce3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016930.2 | NP_001016930.1 | 6 - 7 | 491 | 73 | |
XM_012954225.2 | XP_012809679.1 | 6 - 7 | 597 | 73 | ||
XM_012954226.2 | XP_012809680.1 | 6 - 7 | 524 | 73 | ||
GenBank mRNA | BC155975.1 | - | 6 - 7 | 496 | 73 | |
CR762194.2 | - | 6 - 7 | 491 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |