Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017287.2 | NP_001017287.1 | 2 - 3 | 139 | 102 | |
GenBank mRNA | CR855695.2 | - | 2 - 3 | 139 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |