Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203628.2 | NP_988959.1 | 1 - 2 | 215 | 86 | |
GenBank mRNA | BC061268.1 | - | 1 - 2 | 226 | 86 | |
CR760892.2 | - | 1 - 2 | 215 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |