Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203976.2 | NP_989307.1 | 3 - 4 | 498 | 86 | |
XM_012958392.2 | XP_012813846.1 | 4 - 5 | 606 | 86 | ||
XM_012958402.2 | XP_012813856.1 | 4 - 5 | 584 | 86 | ||
XM_012958408.2 | XP_012813862.1 | 4 - 5 | 584 | 86 | ||
XM_012958414.2 | XP_012813868.1 | 4 - 5 | 584 | 86 | ||
GenBank mRNA | BC064266.1 | - | 3 - 4 | 504 | 86 | |
BC080950.1 | - | 3 - 4 | 502 | 86 | ||
CR760104.2 | - | 3 - 4 | 498 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |