Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015932.3 | NP_001015932.1 | 3 - 4 | 267 | 109 | |
XM_018096248.1 | XP_017951737.1 | 2 - 3 | 236 | 109 | ||
GenBank mRNA | BC122940.1 | - | 3 - 4 | 267 | 109 | |
CR926375.2 | - | 3 - 4 | 263 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |