Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044451.1 | NP_001037916.1 | 4 - 5 | 972 | 78 | |
GenBank mRNA | CR760952.1 | - | 4 - 5 | 972 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |