Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045645.2 | NP_001039110.1 | 3 - 4 | 350 | 103 | |
XM_012957158.2 | XP_012812612.1 | 3 - 4 | 460 | 103 | ||
XM_012957160.2 | XP_012812614.1 | 3 - 4 | 439 | 103 | ||
XM_012957161.2 | XP_012812615.1 | 3 - 4 | 446 | 103 | ||
XM_012957162.2 | XP_012812616.1 | 3 - 4 | 813 | 103 | ||
GenBank mRNA | BC135368.1 | - | 3 - 4 | 357 | 103 | |
CR942425.2 | - | 3 - 4 | 352 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |