Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045663.1 | NP_001039128.1 | 2 - 3 | 134 | 82 | |
GenBank mRNA | CR942486.1 | - | 2 - 3 | 134 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |