Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045786.1 | NP_001039251.1 | 1 - 2 | 438 | 95 | |
GenBank mRNA | CR942794.2 | - | 1 - 2 | 438 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |