Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045800.1 | NP_001039265.1 | 4 - 5 | 295 | 97 | |
XM_012959205.2 | XP_012814659.1 | 4 - 5 | 389 | 97 | ||
GenBank mRNA | CR942742.2 | - | 4 - 5 | 295 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |