Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078786.1 | NP_001072254.1 | 8 - 9 | 1130 | 85 | |
XM_012965019.2 | XP_012820473.1 | 8 - 9 | 1207 | 85 | ||
XM_012965024.2 | XP_012820478.1 | 8 - 9 | 1208 | 85 | ||
XM_018094323.1 | XP_017949812.1 | 5 - 6 | 482 | 85 | ||
GenBank mRNA | BC121219.1 | - | 8 - 9 | 1130 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |