Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079206.1 | NP_001072674.1 | 8 - 9 | 1095 | 78 | |
XM_012957298.2 | XP_012812752.1 | 7 - 8 | 1322 | 78 | ||
GenBank mRNA | BC124001.1 | - | 8 - 9 | 1095 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |