Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079219.1 | NP_001072687.1 | 4 - 5 | 564 | 104 | |
XM_012965802.1 | XP_012821256.1 | 4 - 5 | 619 | 104 | ||
XM_012965806.2 | XP_012821260.1 | 4 - 5 | 622 | 104 | ||
XM_018094605.1 | XP_017950094.1 | 4 - 5 | 313 | 104 | ||
GenBank mRNA | BC124020.1 | - | 4 - 5 | 564 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |