Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079244.1 | NP_001072712.1 | 4 - 5 | 412 | 100 | |
XM_012965828.2 | XP_012821282.1 | 5 - 6 | 659 | 100 | ||
GenBank mRNA | BC124065.1 | - | 4 - 5 | 412 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |