Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097278.1 | NP_001090747.1 | 3 - 4 | 497 | 71 | |
GenBank mRNA | AM419014.1 | - | 3 - 4 | 497 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Xenopus tropicalis Probe spans exons |