Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097378.1 | NP_001090847.1 | 6 - 7 | 832 | 89 | |
GenBank mRNA | BC123095.1 | - | 6 - 7 | 832 | 89 | |
BC158917.1 | - | 6 - 7 | 795 | 89 | ||
CU025106.1 | - | 6 - 7 | 862 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |