Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102704.1 | NP_001096174.1 | 4 - 5 | 967 | 82 | |
XM_012956267.2 | XP_012811721.1 | 4 - 5 | 1303 | 82 | ||
GenBank mRNA | BC136036.1 | - | 4 - 5 | 967 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |