Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102778.1 | NP_001096248.1 | 4 - 5 | 998 | 86 | |
GenBank mRNA | BC135402.1 | - | 4 - 5 | 998 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |