Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112913.1 | NP_001106384.1 | 1 - 2 | 128 | 68 | |
GenBank mRNA | BC155513.1 | - | 1 - 2 | 128 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |