Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113672.1 | NP_001107144.1 | 2 - 3 | 202 | 72 | |
XM_012960554.2 | XP_012816008.1 | 2 - 3 | 560 | 72 | ||
XM_012960555.2 | XP_012816009.1 | 2 - 3 | 560 | 72 | ||
GenBank mRNA | BC155999.1 | - | 2 - 3 | 202 | 72 | |
BC170810.1 | - | 2 - 3 | 195 | 72 | ||
BC170812.1 | - | 2 - 3 | 191 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |