Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123422.1 | NP_001116894.1 | 5 - 6 | 678 | 88 | |
XM_012967973.2 | XP_012823427.1 | 5 - 6 | 729 | 88 | ||
XM_012967974.2 | XP_012823428.1 | 6 - 7 | 843 | 88 | ||
GenBank mRNA | BC159321.1 | - | 5 - 6 | 678 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |