Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123447.1 | NP_001116919.1 | 2 - 3 | 189 | 92 | |
XM_012960584.2 | XP_012816038.1 | 2 - 3 | 280 | 92 | ||
XM_012960585.2 | XP_012816039.1 | 3 - 4 | 385 | 92 | ||
XM_018092916.1 | XP_017948405.1 | 3 - 4 | 387 | 92 | ||
GenBank mRNA | BC161493.1 | - | 2 - 3 | 189 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |