Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126742.1 | NP_001120214.1 | 5 - 6 | 764 | 66 | |
GenBank mRNA | BC159378.1 | - | 5 - 6 | 764 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |