Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: carnmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126522.1 | NP_001119994.1 | 5 - 6 | 1200 | 96 | |
GenBank mRNA | BC158255.1 | - | 5 - 6 | 1200 | 96 | |
CU024918.1 | - | 5 - 6 | 825 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |