Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlycd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126941.1 | NP_001120413.1 | 2 - 3 | 709 | 78 | |
GenBank mRNA | BC161140.1 | - | 2 - 3 | 709 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |