Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130202.1 | NP_001123674.1 | 7 - 8 | 1066 | 81 | |
XM_018091949.1 | XP_017947438.1 | 8 - 9 | 1386 | 81 | ||
GenBank mRNA | BC166965.1 | - | 7 - 8 | 1066 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |