Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134810.1 | NP_001128282.1 | 12 - 13 | 2570 | 84 | |
GenBank mRNA | BC168574.1 | - | 12 - 13 | 2570 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |