Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142023.1 | NP_001135495.1 | 3 - 4 | 427 | 84 | |
XM_012970172.1 | XP_012825626.1 | 3 - 4 | 536 | 84 | ||
GenBank mRNA | BC167895.1 | - | 3 - 4 | 427 | 84 | |
CU075616.1 | - | 3 - 4 | 406 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |