Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: h2afx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015968.2 | NP_001015968.1 | 1 | 348 | 108 | |
GenBank mRNA | CR848416.2 | - | 1 | 348 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |