Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100281180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154099.1 | NP_001147571.1 | 893 | 93 | ||
GenBank mRNA | EU955837.1 | - | 893 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Protease Zea mays |