Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111999.1 | NP_001105469.1 | 737 | 73 | ||
GenBank mRNA | X85803.1 | - | 737 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |