Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111569.1 | NP_001105039.1 | 741 | 99 | ||
GenBank mRNA | AF330036.1 | - | 741 | 99 | ||
AF330037.1 | - | 592 | 99 | |||
BT040253.1 | - | 738 | 99 | |||
BT043171.1 | - | 776 | 99 | |||
EU961678.1 | - | 794 | 99 | |||
L39789.1 | - | 715 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |