Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111945.1 | NP_001105415.1 | 1338 | 88 | ||
GenBank mRNA | X12538.1 | - | 1338 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |