Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332212.1 | NP_001323115.1 | 3 - 4 | 702 | 76 | |
NM_101486.3 | NP_173070.1 | 4 - 5 | 456 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |