Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKDSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035978.2 | NP_001031055.1 | 7 - 8 | 539 | 99 | |
NM_001084078.1 | NP_001077547.1 | 7 - 8 | 479 | 99 | ||
NM_001198082.1 | NP_001185011.1 | 6 - 7 | 615 | 99 | ||
NM_101500.4 | NP_173084.1 | 6 - 7 | 728 | 99 | ||
GenBank mRNA | AK175407.1 | - | 7 - 8 | 511 | 99 | |
BT010438.1 | - | 6 - 7 | 436 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |