Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101814.5 | NP_173387.1 | 2 - 3 | 262 | 98 | |
GenBank mRNA | AK117865.1 | - | 2 - 3 | 190 | 98 | |
AY039590.1 | - | 2 - 3 | 194 | 98 | ||
AY052211.1 | - | 2 - 3 | 168 | 98 | ||
AY055790.1 | - | 2 - 3 | 152 | 98 | ||
AY085426.1 | - | 2 - 3 | 190 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |