Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101938.5 | NP_564126.1 | 1 - 2 | 575 | 57 | |
GenBank mRNA | AF191028.1 | - | 1 - 2 | 483 | 57 | |
AK227749.1 | - | 1 - 2 | 511 | 57 | ||
BT004822.1 | - | 1 - 2 | 478 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |