Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345737.1 | NP_001332639.1 | 17 - 18 | 2256 | 112 | |
NM_001345738.1 | NP_001332640.1 | 17 - 18 | 2231 | 112 | ||
NM_126000.2 | NP_569025.2 | 16 - 17 | 1961 | 112 | ||
NM_180946.3 | NP_851277.1 | 17 - 18 | 2291 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |