Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126036.4 | NP_569032.1 | 9 - 10 | 1057 | 89 | |
GenBank mRNA | AK227464.1 | - | 9 - 10 | 922 | 89 | |
BT010139.1 | - | 9 - 10 | 790 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |