Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126091.4 | NP_569041.1 | 1 - 2 | 429 | 65 | |
GenBank mRNA | AY094401.1 | - | 1 - 2 | 280 | 65 | |
BT000862.1 | - | 1 - 2 | 167 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |