Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126227.3 | NP_565272.1 | 2 - 3 | 1049 | 114 | |
NM_201665.2 | NP_973394.1 | 1 - 2 | 1470 | 114 | ||
GenBank mRNA | AY062817.1 | - | 2 - 3 | 903 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |