Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111240.2 | NP_187019.1 | 3 - 4 | 718 | 71 | |
GenBank mRNA | KJ138842.1 | - | 3 - 4 | 684 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |