Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118347.3 | NP_193953.1 | 2 - 3 | 533 | 69 | |
GenBank mRNA | AF361579.1 | - | 2 - 3 | 407 | 69 | |
AJ866966.1 | - | 2 - 3 | 390 | 69 | ||
AY081721.1 | - | 2 - 3 | 357 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |